Learn More
Cayman Chemical 5-EthynylurIdIn 50mg
SDP
Catalog No. 502947699
Encompass_Preferred
A clickable form of uridine; is incorporated into cellular RNA and has been used to detect transcription in vitro and in vivo
Catalog No. 50-294-7699 Supplier Cayman Chemical Supplier No. 3439150 MG
A clickable form of uridine; is incorporated into cellular RNA and has been used to detect transcription in vitro and in vivo