Learn More
Abcam 5-FAM (5-Carbo x yfluorescein), Amine-reactive fluorescent label, 25MG
Catalog No. 502705318
Encompass_Preferred
MW 376.3 Da, Purity >99%. Amine-reactive fluorescent label. 6-FAM (ab145294) isomer. Polar tracer. Covalently labels peptides and proteins. 5-CFDA N-succinimidyl ester (ab145292) and 5-CFDA (ab145256) hydrolytic product.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5318 Supplier Abcam Supplier No. AB14529325MG
MW 376.3 Da, Purity >99%. Amine-reactive fluorescent label. 6-FAM (ab145294) isomer. Polar tracer. Covalently labels peptides and proteins. 5-CFDA N-succinimidyl ester (ab145292) and 5-CFDA (ab145256) hydrolytic product.
The product is subject to the following: Abcam Restricted Use Statement