Learn More
Medchemexpress LLC 5-fam-ahx-ll-37 tfa | 98.06% | 4964.72 (free acid) | 1 MG
SDP
Catalog No. 5000399955
Encompass_Preferred
5-FAM-Ahx-LL-37 TFA is a 5-FAM-labeled human LL-37, a 37-residue, amphipathic, cathelicidin-derived antimicrobial peptide. The carboxyfluorescein group is attached via a 6-carbon spacer, 6-Aminohexanoic acid. This peptide exhibits a broad spectrum of antimicrobial activity.
  • Appears as a solid
  • Sequence is {5-FAM-Ahx}-Leu-Leu-Gly-Asp-Phe-Phe-Arg-Lys-Ser-Lys-Glu-Lys-Ile-Gly-Lys-Glu-Phe-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-R-Asn-Leu-Val-Pro-Arg-Thr-Glu-Ser
  • Ships at room temperature in continental US
  • Store powder sealed away from moisture at -80°C for 2 years or -20°C for 1 year
  • Store in solvent sealed away from moisture at -80°C for 6 months or -20°C for 1 month
  • Soluble in H2O at 16.67 mg/mL (requires ultrasonic assistance)
  • Classified as an antimicrobial peptide
Catalog No. 50-003-99955 Supplier Medchemexpress LLC Supplier No. HYP5057B1MG
5-FAM-Ahx-LL-37 TFA is a 5-FAM-labeled human LL-37, a 37-residue, amphipathic, cathelicidin-derived antimicrobial peptide. The carboxyfluorescein group is attached via a 6-carbon spacer, 6-Aminohexanoic acid. This peptide exhibits a broad spectrum of antimicrobial activity.
  • Appears as a solid
  • Sequence is {5-FAM-Ahx}-Leu-Leu-Gly-Asp-Phe-Phe-Arg-Lys-Ser-Lys-Glu-Lys-Ile-Gly-Lys-Glu-Phe-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-R-Asn-Leu-Val-Pro-Arg-Thr-Glu-Ser
  • Ships at room temperature in continental US
  • Store powder sealed away from moisture at -80°C for 2 years or -20°C for 1 year
  • Store in solvent sealed away from moisture at -80°C for 6 months or -20°C for 1 month
  • Soluble in H2O at 16.67 mg/mL (requires ultrasonic assistance)
  • Classified as an antimicrobial peptide