Learn More
Medchemexpress LLC 5-FAM-Alkyne | 510758-19-7 | 95.0% | 50 MG
SDP
Catalog No. 5000338938
Encompass_Preferred
5-FAM-Alkyne is a highly selective and sensitive fluorescent biosensor for alkaline phosphatase (ALP). It functions as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. The FAM fluorophore readily diffuses in solution, producing a strong fluorescence signal. This product is intended for research use only.
  • Highly selective and sensitive fluorescent biosensor for alkaline phosphatase (ALP)
  • Functions as a click chemistry reagent
  • Contains an alkyne group for click chemistry reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • FAM fluorophore diffuses easily into solution
  • Emits a strong fluorescence signal
Catalog No. 50-003-38938 Supplier Medchemexpress LLC Supplier No. HY13091350MG
5-FAM-Alkyne is a highly selective and sensitive fluorescent biosensor for alkaline phosphatase (ALP). It functions as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. The FAM fluorophore readily diffuses in solution, producing a strong fluorescence signal. This product is intended for research use only.
  • Highly selective and sensitive fluorescent biosensor for alkaline phosphatase (ALP)
  • Functions as a click chemistry reagent
  • Contains an alkyne group for click chemistry reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • FAM fluorophore diffuses easily into solution
  • Emits a strong fluorescence signal