Learn More
Medchemexpress LLC 5-FAM-alkyne | 510758-19-7 | MFCD28505571 | 95.0% | 413.38 g/mol | C24H15NO6 | 5 MG
SDP
Catalog No. 5000262929
Encompass_Preferred
5-FAM-Alkyne is an alkyne-functionalized 5-carboxyfluorescein fluorescent dye for click-chemistry conjugation and fluorescence-based assays. Supplied as a yellow-to-brown solid with a reported purity of 95.0%, it is intended for research applications requiring covalent labeling of biomolecules.
  • Alkyne-functionalized for click chemistry conjugation.
  • Bright fluorescein-derived dye suitable for fluorescence assays.
  • Yellow-to-brown solid appearance for solid-phase handling.
  • Typical purity 95.0% (verify batch COA for exact value).
  • Molecular formula C24H15NO6 and molecular weight 413.38 g/mol.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-62929 Supplier Medchemexpress LLC Supplier No. HY1309135MG
5-FAM-Alkyne is an alkyne-functionalized 5-carboxyfluorescein fluorescent dye for click-chemistry conjugation and fluorescence-based assays. Supplied as a yellow-to-brown solid with a reported purity of 95.0%, it is intended for research applications requiring covalent labeling of biomolecules.
  • Alkyne-functionalized for click chemistry conjugation.
  • Bright fluorescein-derived dye suitable for fluorescence assays.
  • Yellow-to-brown solid appearance for solid-phase handling.
  • Typical purity 95.0% (verify batch COA for exact value).
  • Molecular formula C24H15NO6 and molecular weight 413.38 g/mol.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months) or -20°C (1 month).