Learn More
Bachem 5-FAM-Amyloid Beta Protein (1-42) Trifluoroacetate
SDP
Supplier:  Bachem 4090151.0500
Encompass_Preferred
5-FAM-Amyloid Beta Protein (1-42) Trifluoroacetate, Fluorescein-5-carbonyl-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, 1802087-78-0, C224H321N55O66S, Mr 4872.41, 0.5mg. 5-FAM-Ab42, b-Amyloid (1-42), FAM - labeled.
Catalog No. 50-194-6617