Learn More
Cayman Chemical 5-FAM-LL-37 (human) (trifluoroacetate salt), Purity: ≥90%, 10 mg
SDP
Catalog No. 502916190
Encompass_Preferred
A fluorescently labeled form of LL-37; ex/em = 492/518 nm
Catalog No. 50-291-6190 Supplier Cayman Chemical Supplier No. 3752410 MG
A fluorescently labeled form of LL-37; ex/em = 492/518 nm