Learn More
Cayman Chemical 5-FAM-l-37humntrIfluor 1mg
SDP
Catalog No. 502917987
Encompass_Preferred
A fluorescently labeled form of LL-37; ex/em = 492/518 nm
Catalog No. 50-291-7987 Supplier Cayman Chemical Supplier No. 375241 MG
A fluorescently labeled form of LL-37; ex/em = 492/518 nm