Learn More
Medchemexpress LLC 5-FAM-LL-37 TFA | 95.76% | 4851.56 (free acid) | 1 MG
SDP
Catalog No. 5000398272
Encompass_Preferred
5-FAM-LL-37 TFA is the TFA salt form of 5-FAM-LL-37, a LL-37 peptide labeled with fluorescein. It retains the antibacterial and immunomodulatory activities of LL-37 by binding to the bacterial cell membrane, thereby destroying its integrity and demonstrating broad-spectrum antibacterial efficacy.
  • Retains antibacterial and immunomodulatory activities
  • Binds to bacterial cell membrane
  • Demonstrates broad-spectrum antibacterial efficacy
Catalog No. 50-003-98272 Supplier Medchemexpress LLC Supplier No. HYP5057A1MG
5-FAM-LL-37 TFA is the TFA salt form of 5-FAM-LL-37, a LL-37 peptide labeled with fluorescein. It retains the antibacterial and immunomodulatory activities of LL-37 by binding to the bacterial cell membrane, thereby destroying its integrity and demonstrating broad-spectrum antibacterial efficacy.
  • Retains antibacterial and immunomodulatory activities
  • Binds to bacterial cell membrane
  • Demonstrates broad-spectrum antibacterial efficacy