Learn More
Medchemexpress LLC HY-15938 10mg Medchemexpress, 5-FAM SE CAS:92557-80-7 Purity:>98%
SDP
Catalog No. 502081841
Encompass_Preferred
Medchemexpress, HY-15938 10mg 5-FAM SE CAS:92557-80-7 5-FAM SE is a single isomer, is a fluorescent labeling reagent used for labeling peptides, proteins and nucleotides. 5-FAM SE can react with amines and can yield stable amine conjugates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1841 Supplier Medchemexpress LLC Supplier No. HY1593810MG
Medchemexpress, HY-15938 10mg 5-FAM SE CAS:92557-80-7 5-FAM SE is a single isomer, is a fluorescent labeling reagent used for labeling peptides, proteins and nucleotides. 5-FAM SE can react with amines and can yield stable amine conjugates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.