Learn More
Aat Bioquest 5-FAM-YVAD-FMK (100 Tests)
SDP
Supplier:  Aat Bioquest 13483
Encompass_Preferred
FAM-YVAD-FMK i.e. 5-carboxyfluorescein-Tyr-Val-Ala-Asp-fluoromethylketone binds irreversibly to active caspase 1 in stimulated cells. This catalog is for 100 Tests.
Catalog No. 50-224-9443