Learn More
Cayman Chemical 5-Feruloylquinic Acid, Purity: ≥98%, CAS: 40242-06-6, 10 mg
SDP
Catalog No. 502946274
Encompass_Preferred
A chlorogenic acid with antioxidant activity; scavenges DPPH and superoxide anion radicals in cell-free assays (IC50s = ~9 and ~36 µM, respectively)
Catalog No. 50-294-6274 Supplier Cayman Chemical Supplier No. 3434910 MG
A chlorogenic acid with antioxidant activity; scavenges DPPH and superoxide anion radicals in cell-free assays (IC50s = ~9 and ~36 µM, respectively)