Learn More
AdipoGen 5-FITC
SDP
Supplier:  AdipoGen CDXF0011M100
Encompass_Preferred
Chemical. CAS 3326-32-7. Formula C21H11NO5S. MW 389.38. Synthetic. Fluorescent marker for biochemical applications. Reacts under mild conditions with primary amines. Used in modification of actin at lys-61, labelling of FAB and the modification of thiol groups.
Catalog No. 50-256-0941