Learn More
Abcam 5-FITC (5-Fluorescein Isothiocyanate), Fluorescent marker, 1G
Catalog No. 502705288
Encompass_Preferred
MW 389.4 Da, Purity >98%. Fluorescent marker. Amine reactive fluorescein isomer. Yields conjugates with superior stability compared to other FITC isomers. Donor for FRET assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5288 Supplier Abcam Supplier No. AB1453251G
MW 389.4 Da, Purity >98%. Fluorescent marker. Amine reactive fluorescein isomer. Yields conjugates with superior stability compared to other FITC isomers. Donor for FRET assays.
The product is subject to the following: Abcam Restricted Use Statement