Learn More
AdipoGen 5-FITC DA
SDP
Supplier:  AdipoGen CDXF0006M050
Encompass_Preferred
Chemical. CAS 118378-76-0. Formula C25H15NO7S. MW 473.45. Synthetic. Fluorescein-diacetate-5-isothiocyanate has a lambdaex of 497nm and a lambdaem of 517nm in 0.1 M phosphate pH 7.0.
Catalog No. 50-256-0946