Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-1-indanone, 99%, Thermo Scientific™

Catalog No. AAL1094906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1094906 5 g
AAL1094903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1094906 Supplier Thermo Scientific Chemicals Supplier No. L1094906

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-84-5
Molecular Formula C9H7FO
Molecular Weight (g/mol) 150.15
MDL Number MFCD00041031
InChI Key WVPPBVAMKNQXJA-UHFFFAOYSA-N
Synonym 5-fluoro-1-indanone, 5-fluoro-2,3-dihydro-1h-inden-1-one, 5-fluoroindan-1-one, 1h-inden-1-one, 5-fluoro-2,3-dihydro, 2,3-dihydro-5-fluoro-1h-inden-1-one, 5-fluoroindanone, 5-fluoro indanone, pubchem2388, 5-flouro-1-indanone, pubchem13785
PubChem CID 136537
IUPAC Name 5-fluoro-2,3-dihydroinden-1-one
SMILES FC1=CC=C2C(=O)CCC2=C1

Specifications

Melting Point 37°C to 40°C
Boiling Point 92°C to 93°C (3 mmHg)
Flash Point 109°C (228°F)
Quantity 5 g
Beilstein 1617799
Formula Weight 150.15
Percent Purity 99%
Chemical Name or Material 5-Fluoro-1-indanone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 615-057-2
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.