Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-1-indanone 96.0+%, TCI America™
SDP

Catalog No. F03761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03761G 1 g
F03765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03761G Supplier TCI America Supplier No. F03761G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 700-84-5
Molecular Formula C9H7FO
Molecular Weight (g/mol) 150.15
MDL Number MFCD00041031
InChI Key WVPPBVAMKNQXJA-UHFFFAOYSA-N
Synonym 5-fluoro-1-indanone, 5-fluoro-2,3-dihydro-1h-inden-1-one, 5-fluoroindan-1-one, 1h-inden-1-one, 5-fluoro-2,3-dihydro, 2,3-dihydro-5-fluoro-1h-inden-1-one, 5-fluoroindanone, 5-fluoro indanone, pubchem2388, 5-flouro-1-indanone, pubchem13785
PubChem CID 136537
IUPAC Name 5-fluoro-2,3-dihydro-1H-inden-1-one
SMILES FC1=CC=C2C(=O)CCC2=C1

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 114°C
Quantity 1 g
Formula Weight 150.15
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-1-indanone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.