Learn More
Medchemexpress LLC 5-fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione | 2230957-36-3 | 290.25 | C14H11FN2O4 | 250 MG
SDP
Catalog No. 5000284291
Encompass_Preferred
Thalidomide-1-Me,5-F is a fluorinated thalidomide derivative used as an E3 ubiquitin ligase activator in research. Supplied as a solid for laboratory use, it functions as a synthetic building block for further derivatization and for studies that probe E3 ligase-mediated pathways. Consult the product datasheet for lot-specific purity and analytical data.
  • Fluorinated E3 ligase activator suitable for biochemical assays
  • Reactive position enables SNAr derivatization for synthesis
  • Supplied as a solid in multiple laboratory scales
  • Molecular formula C14H11FN2O4; molecular weight 290.25
  • Powder storage: -20°C (long term) or 4°C (short term)
  • Data sheet and certificate of analysis available for purity and handling
Catalog No. 50-002-84291 Supplier Medchemexpress LLC Supplier No. HYW577257250MG
Thalidomide-1-Me,5-F is a fluorinated thalidomide derivative used as an E3 ubiquitin ligase activator in research. Supplied as a solid for laboratory use, it functions as a synthetic building block for further derivatization and for studies that probe E3 ligase-mediated pathways. Consult the product datasheet for lot-specific purity and analytical data.
  • Fluorinated E3 ligase activator suitable for biochemical assays
  • Reactive position enables SNAr derivatization for synthesis
  • Supplied as a solid in multiple laboratory scales
  • Molecular formula C14H11FN2O4; molecular weight 290.25
  • Powder storage: -20°C (long term) or 4°C (short term)
  • Data sheet and certificate of analysis available for purity and handling