Learn More
Medchemexpress LLC 5-fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione | 2230957-36-3 | 97.0% | 290.25 g/mol | C14H11FN2O4 | 5 G
SDP
Catalog No. 5000284134
Encompass_Preferred
A fluorinated thalidomide analogue that functions as an E3 ligase (Cereblon) activator and a synthetic building block for targeted protein degradation research. Suitable for recruiting CRBN in PROTAC design and for further chemical derivatization via nucleophilic aromatic substitution (SNAr).
  • Fluorinated thalidomide analogue
  • Acts as an E3 ligase (Cereblon) activator
  • Useful as a PROTAC ligand building block
  • Compatible with SNAr derivatization
  • Typical purity approximately 97.0%
  • Molecular formula C14H11FN2O4; molecular weight 290.25 g/mol
Catalog No. 50-002-84134 Supplier Medchemexpress LLC Supplier No. HYW5772575G
A fluorinated thalidomide analogue that functions as an E3 ligase (Cereblon) activator and a synthetic building block for targeted protein degradation research. Suitable for recruiting CRBN in PROTAC design and for further chemical derivatization via nucleophilic aromatic substitution (SNAr).
  • Fluorinated thalidomide analogue
  • Acts as an E3 ligase (Cereblon) activator
  • Useful as a PROTAC ligand building block
  • Compatible with SNAr derivatization
  • Typical purity approximately 97.0%
  • Molecular formula C14H11FN2O4; molecular weight 290.25 g/mol