Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione | 2230957-36-3 | >98.0% | 290.25 | C14H11FN2O4 | 1 G
SDP

Catalog No. 5000283878
Encompass_Preferred

Thalidomide-1-Me,5-F is a fluorinated thalidomide derivative (5-fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione) described as an E3 ligase activator. It is provided as a research chemical for use as a biochemical probe, synthetic building block, or starting material for further derivatization.

  • Acts as an E3 ligase activator useful in targeted protein degradation studies.
  • Contains a fluorine atom enabling SNAr reactions and further derivatization.
  • Supplied in small-quantity packages suitable for research and synthesis.
  • Well-defined chemical identity with CAS 2230957-36-3.
  • Chemical formula C14H11FN2O4 and molecular weight 290.25.

Catalog No. 50-002-83878 Supplier Medchemexpress LLC Supplier No. HYW5772571G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-1-Me,5-F is a fluorinated thalidomide derivative (5-fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione) described as an E3 ligase activator. It is provided as a research chemical for use as a biochemical probe, synthetic building block, or starting material for further derivatization.

  • Acts as an E3 ligase activator useful in targeted protein degradation studies.
  • Contains a fluorine atom enabling SNAr reactions and further derivatization.
  • Supplied in small-quantity packages suitable for research and synthesis.
  • Well-defined chemical identity with CAS 2230957-36-3.
  • Chemical formula C14H11FN2O4 and molecular weight 290.25.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.