Learn More
Medchemexpress LLC 5-fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione | 2230957-36-3 | MFCD30188075 | 99.9% | 276.22 g/mol | C13H9FN2O4 | 100 MG
SDP
Catalog No. 5000284100
Encompass_Preferred
5-Fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione is a fluorinated thalidomide derivative that functions as an E3 ligase (cereblon) ligand. It is provided as a high-purity solid and is intended for use as a chemical building block in targeted protein degradation research and for further derivatization via nucleophilic aromatic substitution (SNAr).
  • Functions as an E3 ligase (cereblon) ligand.
  • Useful as a building block for PROTAC and targeted protein degradation design.
  • Fluorinated position enables SNAr-based further derivatization.
  • Supplied as a high-purity small-molecule solid suitable for synthetic chemistry.
  • Molecular formula C13H9FN2O4; molecular weight 276.22 g/mol.
Catalog No. 50-002-84100 Supplier Medchemexpress LLC Supplier No. HYW577257100MG
5-Fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione is a fluorinated thalidomide derivative that functions as an E3 ligase (cereblon) ligand. It is provided as a high-purity solid and is intended for use as a chemical building block in targeted protein degradation research and for further derivatization via nucleophilic aromatic substitution (SNAr).
  • Functions as an E3 ligase (cereblon) ligand.
  • Useful as a building block for PROTAC and targeted protein degradation design.
  • Fluorinated position enables SNAr-based further derivatization.
  • Supplied as a high-purity small-molecule solid suitable for synthetic chemistry.
  • Molecular formula C13H9FN2O4; molecular weight 276.22 g/mol.