Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Fluoro-2 -deoxycyt 10mM 1mL | 10356-76-0 | 245.21 g/mol | C9H12FN3O4 | 1 ML
SDP

Catalog No. 5000228376
Encompass_Preferred

5-Fluoro-2'-deoxycytidine is a fluorinated deoxycytidine analogue that functions as a DNA methyltransferase (DNMT) inhibitor. Supplied as a prepared 10 mM solution (1 mL), it is intended for research use in epigenetics, DNA methylation studies, and cancer biology.

  • Prepared solution at 10 mM concentration.
  • Supplied as 1 mL volume, ready for immediate use.
  • High purity (99.66% by HPLC) as reported by supplier COA.
  • Molecular formula C9H12FN3O4; molecular weight 245.21 g/mol.
  • Intended for in vitro research; not for human or clinical use.
  • Store as recommended by supplier (typically -20°C) to preserve stability.

Catalog No. 50-002-28376 Supplier Medchemexpress LLC Supplier No. HY11621710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

5-Fluoro-2'-deoxycytidine is a fluorinated deoxycytidine analogue that functions as a DNA methyltransferase (DNMT) inhibitor. Supplied as a prepared 10 mM solution (1 mL), it is intended for research use in epigenetics, DNA methylation studies, and cancer biology.

  • Prepared solution at 10 mM concentration.
  • Supplied as 1 mL volume, ready for immediate use.
  • High purity (99.66% by HPLC) as reported by supplier COA.
  • Molecular formula C9H12FN3O4; molecular weight 245.21 g/mol.
  • Intended for in vitro research; not for human or clinical use.
  • Store as recommended by supplier (typically -20°C) to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.