Learn More
TARGETMOL CHEMICALS INC 5-FLUORO-2-DEOXYCYTIDINE 50MG
SDP
Catalog No. 502579158
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5-Fluoro-2'-deoxycytidine (2'-DEOXY-5-FLUOROCYTIDINE) is an inhibitor of DNA methyltransferase (DNMT) . purity: 99%
Catalog No. 50-257-9158 Supplier TARGETMOL CHEMICALS INC Supplier No. T771850MG
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5-Fluoro-2'-deoxycytidine (2'-DEOXY-5-FLUOROCYTIDINE) is an inhibitor of DNA methyltransferase (DNMT) . purity: 99%