Learn More
Medchemexpress LLC 5-fluoro-2'-deoxyuridine-5'-triphosphate | 2710-64-7 | >95.0% | 486.13 g/mol | C9H14FN2O14P3 | 10 MG
SDP
Catalog No. 5000285872
Encompass_Preferred
5-Fluoro-2'-deoxy-UTP is a fluorinated deoxyuridine triphosphate designed for use as a substrate in enzymatic DNA synthesis and incorporation studies. The molecule carries a fluorine substituent at the 5-position of the uracil base and is supplied as a solid with accompanying safety and technical documentation for laboratory handling.
  • Modified nucleotide triphosphate for enzymatic incorporation.
  • Enables incorporation studies and DNA polymerase assay development.
  • Supplied as a white to off-white solid with SDS and data sheet available.
  • Chemical formula C9H14FN2O14P3 and molecular weight 486.13 g/mol for reference.
  • Available in small pack sizes suitable for research workflows.
Catalog No. 50-002-85872 Supplier Medchemexpress LLC Supplier No. HY15587810MG
5-Fluoro-2'-deoxy-UTP is a fluorinated deoxyuridine triphosphate designed for use as a substrate in enzymatic DNA synthesis and incorporation studies. The molecule carries a fluorine substituent at the 5-position of the uracil base and is supplied as a solid with accompanying safety and technical documentation for laboratory handling.
  • Modified nucleotide triphosphate for enzymatic incorporation.
  • Enables incorporation studies and DNA polymerase assay development.
  • Supplied as a white to off-white solid with SDS and data sheet available.
  • Chemical formula C9H14FN2O14P3 and molecular weight 486.13 g/mol for reference.
  • Available in small pack sizes suitable for research workflows.