Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 5-Fluoro-2'-deoxyuridine, 50-91-9, MFCD00006530, 1g

Catalog No. 501472648
Encompass_Preferred

Molecular Formula C9H11FN2O5, Purity 99%,Molecular Weight 246.20

Catalog No. 50-147-2648 Supplier Matrix Scientific Supplier No. 0089451G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C9H11FN2O5, Purity 99%,Molecular Weight 246.20

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.