Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F09771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09771G 1 g
F0977-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09771G Supplier TCI America Supplier No. F09771G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 5-Fluoro-2-hydroxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 259209-20-6
Melting Point 166°C
Color White-Yellow
Molecular Formula C6H6BFO3
Quantity 1 g
MDL Number MFCD03095357
Synonym 5-fluoro-2-hydroxyphenyl boronic acid, 5-fluoro-2-hydroxybenzeneboronic acid, 2-borono-4-fluorophenol, boronic acid, 5-fluoro-2-hydroxyphenyl, 5-fluoro-2-hydroxy phenylboronic acid, boronic acid,b-5-fluoro-2-hydroxyphenyl, pubchem19871, 5-fluoro-2-hydroxy-phenyl boronic acid, acmc-209gna, ablock ab-12-3331
InChI Key TWDQSJDFXUMAOI-UHFFFAOYSA-N
SMILES OB(O)C1=C(O)C=CC(F)=C1
IUPAC Name (5-fluoro-2-hydroxyphenyl)boronic acid
Molecular Weight (g/mol) 155.92
PubChem CID 3772476
Formula Weight 155.92
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.