Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Fluoro-2-(isoxazol-5-yl)phenol

Catalog No. JRD08491G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD08491G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD08491G Supplier Sigma Aldrich Supplier No. JRD08491G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyl(4-iodobutoxy)dimethylsilane is an iodoorganosilane that can be synthesized from tert-butyl(chloro)dimethylsilane.tert-Butyl(4-iodobutoxy)dimethylsilane may be used as a reagent in the multi-step synthesis of spermine synthon and decanucleotide-spermine conjugates.

Specifications

Molecular Formula C9H6FNO2
Linear Formula C9H6FNO2
MDL Number MFCD12196362
Quantity 25 mL
Molecular Weight (g/mol) 179.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.