Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-methoxybenzonitrile 98.0+%, TCI America™
SDP

Catalog No. F04795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04795G 5 g
F04791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04795G Supplier TCI America Supplier No. F04795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 189628-38-4
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD02683503
InChI Key XNCVSNKXADJGOG-UHFFFAOYSA-N
PubChem CID 2782746
IUPAC Name 5-fluoro-2-methoxybenzonitrile
SMILES COC1=C(C=C(C=C1)F)C#N

Specifications

Color Yellow
Quantity 5 g
Synonym 2-cyano-4-fluoroanisole, 3-fluoro-6-methoxybenzonitrile, benzonitrile, 5-fluoro-2-methoxy, 5-fluoro-2-methoxybenzenecarbonitrile, pubchem3502, acmc-1bxaq, xncvsnkxadjgog-uhfffaoysa, benzonitrile,5-fluoro-2-methoxy, buttpark 80\01-23, 5-fluoro-2-methyloxy benzonitrile
Formula Weight 151.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-methoxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.