Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-methylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. F07475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07475G 5 g
F074725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07475G Supplier TCI America Supplier No. F07475G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77532-79-7
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00042295
InChI Key IBRODYNXELBTJC-UHFFFAOYSA-N
Synonym 2-cyano-4-fluorotoluene, 2-methyl-5-fluorobenzonitrile, benzonitrile, 5-fluoro-2-methyl, 3-fluoro-6-methylbenzonitrile, 5-fluoro-2-methyl-benzonitrile, 5-fluoro-o-tolunitrile, 5-fluoro-2-methylbenzenecarbonitrile, 2-methy-5-fluoro benzonitrile, pubchem1551, acmc-209pak
PubChem CID 522668
IUPAC Name 5-fluoro-2-methylbenzonitrile
SMILES CC1=C(C=C(C=C1)F)C#N

Specifications

Melting Point 43°C
Quantity 5 g
UN Number 3439
Formula Weight 135.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-methylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.