Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-methylbenzyl Bromide 97.0+%, TCI America™
SDP

Catalog No. F07481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07481G 1 g
F07485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07481G Supplier TCI America Supplier No. F07481G

Chemical Identifiers

CAS 261951-71-7
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD01631528
InChI Key DRIYGEORZARQGO-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylbenzyl bromide, 2-bromomethyl-4-fluoro-1-methylbenzene, 2-methyl-5-fluorobenzyl bromide, 5-fluoro-2-methylbenzylbromide, alpha-bromo-4-fluoro-o-xylene, 2-bromomethyl-4-fluorotoluene, 2-bromomethyl-4-fluoro-1-methyl-benzene, benzene, 2-bromomethyl-4-fluoro-1-methyl, pubchem4927
PubChem CID 2774605
IUPAC Name 2-(bromomethyl)-4-fluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)CBr

Specifications

Color Yellow
Boiling Point 86°C
Quantity 1 g
UN Number 3265
Formula Weight 203.05
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Fluoro-2-methylbenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.