Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-fluoro-2-phenyl-1,2-benzothiazol-3-one | 1306638-12-9 | 99.9% | 245.27 g/mol | C13H8FNOS | 5 MG
SDP

Catalog No. 5000221081
Encompass_Preferred

ML089 is a small-molecule research inhibitor of phosphomannose isomerase (PMI), reported to have an IC50 of approximately 1.3 μM. The compound is supplied as a high-purity solid intended for laboratory research and is typically provided in small milligram quantities. Handle with appropriate laboratory precautions and dissolve using suitable solvents such as DMSO.

  • Potent, selective PMI inhibitor with reported IC50 ~1.3 μM.
  • High purity (>99.8%) suitable for biochemical and cellular studies.
  • Molecular weight 245.27 g/mol; formula C13H8FNOS.
  • Soluble in DMSO for assay and formulation preparation.
  • Available in small milligram pack sizes for screening and optimization.

Catalog No. 50-002-21081 Supplier Medchemexpress LLC Supplier No. HY1388025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML089 is a small-molecule research inhibitor of phosphomannose isomerase (PMI), reported to have an IC50 of approximately 1.3 μM. The compound is supplied as a high-purity solid intended for laboratory research and is typically provided in small milligram quantities. Handle with appropriate laboratory precautions and dissolve using suitable solvents such as DMSO.

  • Potent, selective PMI inhibitor with reported IC50 ~1.3 μM.
  • High purity (>99.8%) suitable for biochemical and cellular studies.
  • Molecular weight 245.27 g/mol; formula C13H8FNOS.
  • Soluble in DMSO for assay and formulation preparation.
  • Available in small milligram pack sizes for screening and optimization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.