Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-pyridinecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. F08385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08385G 5 g
F08381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08385G Supplier TCI America Supplier No. F08385G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 107504-08-5 | C6H4FNO2 | 141.101 g/mol

Chemical Identifiers

CAS 107504-08-5
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD04114196
InChI Key JTKFIIQGMVKDNZ-UHFFFAOYSA-N
Synonym 5-fluoropicolinic acid, 5-fluoro-2-picolinic acid, 5-fluoro-2-pyridinecarboxylic acid, 5-fluoropyridine-2-carboxylicacid, 5-fluoro-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-fluoro, 2-carboxy-5-fluoropyridine, 5-fluoro-2-carboxypyridine, pubchem14387, acmc-2098wr
PubChem CID 2762876
IUPAC Name 5-fluoropyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1F)C(=O)O

Specifications

Melting Point 165°C
Quantity 5 g
Formula Weight 141.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-pyridinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.