Learn More
Cayman Chemical 5-fluoro 203, Purity: ≥98%, CAS: 260443-89-8, 10 mg
SDP
Catalog No. 502979555
Encompass_Preferred
A cytotoxic compound that at 1 µM activates aryl hydrocarbon receptor signaling, inducing transcription of cytochrome P450 1A1, which leads to the formation of DNA adducts and cell cycle arrest; can increase the levels of ROS as well as activate JNK, ERK, and p38MAPK in certain ovarian, breast, kidney, and colorectal cancer cells
Catalog No. 50-297-9555 Supplier Cayman Chemical Supplier No. 1767710 MG
A cytotoxic compound that at 1 µM activates aryl hydrocarbon receptor signaling, inducing transcription of cytochrome P450 1A1, which leads to the formation of DNA adducts and cell cycle arrest; can increase the levels of ROS as well as activate JNK, ERK, and p38MAPK in certain ovarian, breast, kidney, and colorectal cancer cells