Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-5'-deoxyuridine, Thermo Scientific Chemicals

Catalog No. AC462790050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462790050 5 g
AC462790010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462790050 Supplier Thermo Scientific Chemicals Supplier No. 462790050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9-5-3094
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD00866530 MFCD00866530
InChI Key ZWAOHEXOSAUJHY-ZIYNGMLESA-N
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

Melting Point 188.0°C to 192.0°C
Quantity 5 g
Formula Weight 246.19
Chemical Name or Material 5-Fluoro-5 '-deoxyuridine
EINECSNumber 221-440-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.