Learn More
Cayman Chemical 5-fluoro ADBICA, Purity: ≥98%, CAS: 1863065-82-0, 10 mg
SDP
Catalog No. 502968566
Encompass_Preferred
A derivative of ADBICA featuring a fluorine atom added to the terminal carbon of the pentyl group; intended for forensic and research applications
Catalog No. 50-296-8566 Supplier Cayman Chemical Supplier No. 1476610 MG
A derivative of ADBICA featuring a fluorine atom added to the terminal carbon of the pentyl group; intended for forensic and research applications