Learn More
Cayman Chemical 5-fluoro MDMB-PICA metabolite
SDP
Supplier:  Cayman Chemical 274881 MG
Encompass_Preferred
An internal standard for the quantification of 5-fluoro MDMB-PICA metabolite 7 by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-294-1496