Learn More
Cayman Chemical 5-fluoro MDMB-PICA metabolite 9, Purity: ≥98%, 1 mg
SDP
Catalog No. 502941622
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-294-1622 Supplier Cayman Chemical Supplier No. 275871 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications