Learn More
Cayman Chemical 5-fluoro MDMB-PICA metabolite 2, Purity: ≥98%, 1 mg
SDP
Catalog No. 502986833
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of 5-fluoro MDMB-PICA and MDMB-PICA; intended for research and forensic applications
Catalog No. 50-298-6833 Supplier Cayman Chemical Supplier No. 223931 MG
An analytical reference standard categorized as a synthetic cannabinoid metabolite; a metabolite of 5-fluoro MDMB-PICA and MDMB-PICA; intended for research and forensic applications