Learn More
Cayman Chemical 5-fluoro MPP-PICA phenylpropi
SDP
Supplier:  Cayman Chemical 312715 MG
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential metabolite of 5-fluoro MPP-PICA based on the published metabolism of 5-fluoro AMB-PICA; intended for research and forensic applications
Catalog No. 50-297-8962