Learn More
Cayman Chemical 5-fluoro nI 2-naphthyl Iso 5mg
SDP
Catalog No. 502953027
Encompass_Preferred
A derivative of NNEI with a fluorine atom at the terminal carbon of the pentyl chain and the naphthyl group joined at the 2 rather than 1 position; intended for forensic and research applications
Catalog No. 50-295-3027 Supplier Cayman Chemical Supplier No. 143185 MG
A derivative of NNEI with a fluorine atom at the terminal carbon of the pentyl chain and the naphthyl group joined at the 2 rather than 1 position; intended for forensic and research applications