Learn More
Cayman Chemical 5-fluoro NNEI, Purity: ≥98%, CAS: 1445580-60-8, 5 mg
SDP
Catalog No. 502953717
Encompass_Preferred
A derivative of NNEI in which a fluorine atom has been added to the terminal carbon of the pentyl chain; intended for forensic and research applications
Catalog No. 50-295-3717 Supplier Cayman Chemical Supplier No. 141475 MG
A derivative of NNEI in which a fluorine atom has been added to the terminal carbon of the pentyl chain; intended for forensic and research applications