Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-o-cresol 98.0+%, TCI America™
SDP

Catalog No. F06321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06321G 1 g
F06325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06321G Supplier TCI America Supplier No. F06321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 452-85-7
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00673009
InChI Key CKJOSIHYVLHIER-UHFFFAOYSA-N
Synonym 4-fluoro-2-hydroxytoluene, 5-fluoro-o-cresol, 5-fluoro-2-methyl-phenol, pubchem1502, 2-methyl-5-fluorophenol, acmc-1afs5, 5-fluoranyl-2-methyl-phenol, ksc494a2f
PubChem CID 2774616
IUPAC Name 5-fluoro-2-methylphenol
SMILES CC1=C(C=C(C=C1)F)O

Specifications

Boiling Point 66°C
Quantity 1 g
Formula Weight 126.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Fluoro-o-cresol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.