Learn More
Cayman Chemical 5-fluoro Orotic Acid (hydrate), Purity: ≥95%, CAS: 220141-70-8, 1 g
SDP
Catalog No. 502967201
Encompass_Preferred
Used to detect URA3 gene expression in yeast molecular genetic constructs
Catalog No. 50-296-7201 Supplier Cayman Chemical Supplier No. 173181 G
Used to detect URA3 gene expression in yeast molecular genetic constructs