Learn More
Cayman Chemical 5-fluoro SDB-005, Purity: ≥98%, CAS: 2185863-14-1, 10 mg
SDP
Catalog No. 502969117
Encompass_Preferred
A derivative of SDB-005 that has a fluorine atom at the terminal carbon of the pentyl chain; intended for research and forensic applications
Catalog No. 50-296-9117 Supplier Cayman Chemical Supplier No. 1535310 MG
A derivative of SDB-005 that has a fluorine atom at the terminal carbon of the pentyl chain; intended for research and forensic applications