Learn More
Cayman Chemical 5-fluoro THJ, Purity: ≥98%, CAS: 2180934-72-7, 10 mg
SDP
Catalog No. 502972290
Encompass_Preferred
A derivative of THJ 018 featuring a fluorine atom added to the terminal carbon of the pentyl group and an 8-aminoquinoline replacing the naphthalene group; intended for research and forensic applications
Catalog No. 50-297-2290 Supplier Cayman Chemical Supplier No. 1489010 MG
A derivative of THJ 018 featuring a fluorine atom added to the terminal carbon of the pentyl group and an 8-aminoquinoline replacing the naphthalene group; intended for research and forensic applications