Learn More
Medchemexpress LLC 5-fluorocytidine | 2341-22-2 | MFCD00210953 | 97.0% | 261.21 g/mol | C9H12FN3O5 | 500 MG
SDP
Catalog No. 5000299477
Encompass_Preferred
5-Fluorocytidine is a fluorinated cytidine analogue used as a research reagent that inhibits maturation of the 45S ribosomal RNA precursor. It is provided as a purified nucleoside for in vitro biochemical and cellular studies; handle according to the SDS and store cold to maintain stability.
  • Inhibits maturation of the 45S ribosomal RNA precursor, useful for rRNA biology studies.
  • Fluorinated nucleoside structure for mechanism-of-action and metabolic investigations.
  • Supplied with batch-specific certificate of analysis for identity and purity.
  • Available in multiple package sizes to support assay screening and scale-up.
  • Stable under recommended cold storage; consult the SDS for handling and storage conditions.
  • Suitable for in vitro biochemical and cell-based assays.
Catalog No. 50-002-99477 Supplier Medchemexpress LLC Supplier No. HYW039722500MG
5-Fluorocytidine is a fluorinated cytidine analogue used as a research reagent that inhibits maturation of the 45S ribosomal RNA precursor. It is provided as a purified nucleoside for in vitro biochemical and cellular studies; handle according to the SDS and store cold to maintain stability.
  • Inhibits maturation of the 45S ribosomal RNA precursor, useful for rRNA biology studies.
  • Fluorinated nucleoside structure for mechanism-of-action and metabolic investigations.
  • Supplied with batch-specific certificate of analysis for identity and purity.
  • Available in multiple package sizes to support assay screening and scale-up.
  • Stable under recommended cold storage; consult the SDS for handling and storage conditions.
  • Suitable for in vitro biochemical and cell-based assays.