Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-fluorocytidine | 2341-22-2 | MFCD00210953 | >98.0% | 261.21 g/mol | C9H12FN3O5 | 1 ML
SDP

Catalog No. 5000299476
Encompass_Preferred

5-fluorocytidine is a fluorinated cytidine nucleoside analogue that interferes with ribosomal RNA processing by inhibiting maturation of the 45S rRNA precursor. It is used as a biochemical tool in cellular and molecular biology research and is available in solid form or as a ready-to-use solution for laboratory assays.

  • Inhibits maturation of the 45S ribosomal RNA precursor.
  • Available as a 10 mM solution in DMSO for ready-to-use assays.
  • High purity (>98% by HPLC) suitable for research applications.
  • Molecular weight 261.21 g/mol and formula C9H12FN3O5 for easy identification.
  • Soluble in DMSO, enabling straightforward formulation for in vitro studies.

Catalog No. 50-002-99476 Supplier Medchemexpress LLC Supplier No. HYW03972210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-fluorocytidine is a fluorinated cytidine nucleoside analogue that interferes with ribosomal RNA processing by inhibiting maturation of the 45S rRNA precursor. It is used as a biochemical tool in cellular and molecular biology research and is available in solid form or as a ready-to-use solution for laboratory assays.

  • Inhibits maturation of the 45S ribosomal RNA precursor.
  • Available as a 10 mM solution in DMSO for ready-to-use assays.
  • High purity (>98% by HPLC) suitable for research applications.
  • Molecular weight 261.21 g/mol and formula C9H12FN3O5 for easy identification.
  • Soluble in DMSO, enabling straightforward formulation for in vitro studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.