Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-fluorocytidine | 2341-22-2 | MFCD00210953 | 99.9% | 261.21 g/mol | C9H12FN3O5 | 100 MG
SDP

Catalog No. 5000300535
Encompass_Preferred

5-Fluorocytidine is a fluorinated cytidine nucleoside analog that inhibits maturation of the 45S ribosomal RNA precursor. It is used in biochemical and cellular research to probe RNA processing and nucleoside metabolism; analytical data and certificates of analysis are available for batch validation.

  • Inhibits maturation of the 45S ribosomal RNA precursor.
  • Molecular formula C9H12FN3O5 and molecular weight 261.21 g/mol.
  • High purity (>99.9%) as supplied; certificate of analysis available.
  • Analytical data available: COA, HNMR, RP-HPLC.
  • Storage: 4°C under nitrogen; in solution, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-00535 Supplier Medchemexpress LLC Supplier No. HYW039722100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Fluorocytidine is a fluorinated cytidine nucleoside analog that inhibits maturation of the 45S ribosomal RNA precursor. It is used in biochemical and cellular research to probe RNA processing and nucleoside metabolism; analytical data and certificates of analysis are available for batch validation.

  • Inhibits maturation of the 45S ribosomal RNA precursor.
  • Molecular formula C9H12FN3O5 and molecular weight 261.21 g/mol.
  • High purity (>99.9%) as supplied; certificate of analysis available.
  • Analytical data available: COA, HNMR, RP-HPLC.
  • Storage: 4°C under nitrogen; in solution, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.