Learn More
Abcam 5-Fluorocytosine (5-FC), antifungal agent, 1G
Catalog No. 502706811
Encompass_Preferred
MW 129.09 Da, Purity >99%. Antimycotic prodrug, converted to 5-fluorouracil. Disrupts DNA and RNA synthesis. Displays antifungal and antitumor activity. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6811 Supplier Abcam Supplier No. AB1411971G
MW 129.09 Da, Purity >99%. Antimycotic prodrug, converted to 5-fluorouracil. Disrupts DNA and RNA synthesis. Displays antifungal and antitumor activity. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement