Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluorodeoxyuridine MP Biomedicals

Catalog No. ICN10555110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10555110 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10555110 Supplier MP Biomedicals, Inc Supplier No. 0210555110
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50-91-9
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD00006530
InChI Key ODKNJVUHOIMIIZ-RRKCRQDMSA-N
Synonym floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
PubChem CID 5790
ChEBI CHEBI:60761
IUPAC Name 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

Quantity 10 mg
Formula Weight 246.2
Chemical Name or Material 5-Fluorodeoxyuridine, Inhibitor of DNA Synthesis.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.